Learn More
Nickel Electro™ Laboratory Stainless Steel Crucible Lid
Shop All Nickel-Electro Products
Click to view available options
Diameter (Metric) Outer:
30 mm
35 mm
40 mm
50 mm
Unit Size:
Pack of 10
Description
Includes
DiameterMetricOuter
Requires
Autoclavable,dimMaterial,ProductTypeSpecs,Quantity,Shape
- Non-magnetic
- Stamped with the word Stainless Steel to assist in the identification of material when in use
- Flat shape with a depression for centering on crucibles
- Includes an extension on one side that serves as a handle
- Crevice free
Non magnetic, bright finish, crevice free lid, handle
Specifications
Specifications
| Material | Stainless Steel |
| Quantity | 10 Pk. |
| Autoclavable | Autoclavable |
| Diameter (Metric) Outer | 30 mm |
| Product Type | Crucible lid |
| Shape | Flat |