Learn More
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 100 L with 3 Ports, 2D BPC
Shop All Thermo Scientific ProductsDescription
Using Thermo Scientific™ Productainer™ BPC bag with 3 ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.
Highlights:
- Three sizes - 50, 100 and 200L
- Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
Recommended for:
Harvest - Collection and storage of bulk product from bioreactors
Storage - Sterile storage of media, buffers and process intermediates
Separation - Feeding and receiving of sterile liquids from separation systems
Purification - Buffer feeding to purification system, fraction collection and storage
Filling - Sterile reservoir for filling systems
Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Specifications
| Film Description | CX5-14 |
| Port Detail | Panel Ports: 2 x 0.38 in. 1 x 0.25 in. |
| For Use With (Equipment) | SV50076.03 |
| Dimensions | Chamber: 71.12 cm x 76.2 cm (28 in. x 30 in.) |
| Material | Polyethylene |
| Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
| Quantity | 100 L, each |
| Sterilization Method | Irradiated, Gamma Irradiated |
| Tubing Line 1 | Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.