missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LACE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556370
This item is not returnable.
View return policy
Description
Immunogen sequence: WKAYTVQTSE SMTPTATSET YLKALAVCHG PLDHYDFLIK AHELKDDEHQ RRVIQCLQKL HEDLKGYNIE AEGLFSKLFS RSKPPRGLYV YGDVGTGK Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 26%.
LACE1 encodes a protein with possible ATPase function. The protein contains a P-loop motif and a five-domain structure that is conserved in fly, yeast, and bacteria. Two conserved estrogen receptor binding sites are located within 2.5 kb of this gene.
Specifications
| LACE1 | |
| Polyclonal | |
| Unconjugated | |
| AFG1L | |
| AFG1; Afg1l; AFG1-like ATPase; ATPase family gene 1 homolog; c222389; CG8520; CG8520 gene product; Lace1; lactation elevated 1; lactation elevated protein 1; Protein AFG1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 246269 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WV93 | |
| AFG1L | |
| Recombinant protein corresponding to Human LACE1. Recombinant protein control fragment (Product #RP-94848). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction