missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACRT Polyclonal Antibody

LACRT Polyclonal Antibody
Brand: Invitrogen PA589952
This item is not returnable.
View return policy
Description
LACRT Polyclonal Antibody for Western Blot
Specifications
| LACRT | |
| Polyclonal | |
| Unconjugated | |
| Extracellular glycoprotein lacritin; lacritin; LACRT | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LACRT (NP_1505931). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction