missing translation for 'onlineSavingsMsg'
Learn More

Lactase Antibody (CL14164) [DyLight 594], Novus Biologicals Biologicals™

Product Code. 30531008 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30531008 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30531008 Supplier Novus Biologicals Supplier No. NBP344274DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lactase
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL14164
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P09848, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3938
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.