missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06457MFV450
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase Polyclonal antibody specifically detects Lactate Dehydrogenase in P. falciparum samples. It is validated for ELISA
Specifications
| Lactate Dehydrogenase | |
| Polyclonal | |
| ELISA | |
| EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
| Produced in rabbits immunized with purified, recombinant Lactate Dehydrogenase (Accession#: XP_001349989.1; Met1-Ala316). | |
| 0.1 mL | |
| HIF Target Genes, Hypoxia, Tumor Biomarkers | |
| 3945 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| P. falciparum | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction