missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lactate Dehydrogenase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30578
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase Polyclonal antibody specifically detects Lactate Dehydrogenase in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Lactate Dehydrogenase | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
| Between 284 and 334 | |
| 100 μg | |
| HIF Target Genes, Hypoxia, Tumor Biomarkers | |
| 3945 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction