missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase B Antibody (2057D) [HRP], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9205H
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase B Monoclonal antibody specifically detects Lactate Dehydrogenase B in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Gene Knock-Out
Specifications
| Lactate Dehydrogenase B | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Gene Knock-Out | |
| 2057D | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated | |
| EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
| E. coli-derived recombinant human LDHB, Met1-Val93, Accession # P07195 | |
| 0.1 mL | |
| HIF Target Genes, Hypoxia, Mitochondrial Mediated Pathway | |
| 3945 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction