missing translation for 'onlineSavingsMsg'
Learn More

LACTB2 Monoclonal Antibody (OTI4E7), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA502583

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17587261

  • € 540.87 / 100µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.
Specifications

Specifications

LACTB2
Monoclonal
0.43 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q53H82
LACTB2
Full length human recombit protein of human LACTB2 produced in E.coli
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI4E7
Unconjugated
LACTB2
CGI-83
Mouse
Affinity Chromatography
RUO
51110
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers