missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Antibody (002), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90704AF532
This item is not returnable.
View return policy
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Mouse samples. It is validated for ELISA.
Specifications
| Lactoferrin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Lactoferrin (AAH06904.1; Met1-Gln707). | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules | |
| 4057 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 2 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction