missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Antibody (1C6cc), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10109IR
This item is not returnable.
View return policy
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA.
Specifications
| Lactoferrin | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LTF | |
| Full length native protein (purified) (Human). | |
| 0.2 mL | |
| Amino Acids Drugs and other small molecules | |
| 4057 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1C6cc | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human lactoferrin. No cross-reactivity with human transferrin. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction