missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Antibody (LF5-1D2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-67260AF532
This item is not returnable.
View return policy
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Flow Cytometry, ELISA, Flow (Intracellular).
Specifications
| Lactoferrin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LTF | |
| Human lactoferrin | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-61 recognizes an epitope on second extracellular domain (EC2) of CD9 antigen, a 24 kDa transmembrane protein expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes. | |
| Store at 4°C in the dark. |
| Flow Cytometry, ELISA | |
| LF5-1D2 | |
| Flow Cytometry, ELISA, Flow (Intracellular) | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| Mouse | |
| Protein A purified | |
| Amino Acids Drugs and other small molecules | |
| 4057.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction