missing translation for 'onlineSavingsMsg'
Learn More

Lactoferrin Antibody (SR2218), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-21725-100ul

 View more versions of this product

Product Code. 18624054

  • € 449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lactoferrin Monoclonal antibody specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lactoferrin
Monoclonal
Unconjugated
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunoprecipitation
SR2218
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
A synthesized peptide derived from human Lactoferrin (Uniprot #: P02788)
100 μg
Amino Acids Drugs and other small molecules
4057
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.