missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lactoferrin Recombinant Rabbit Monoclonal Antibody (103), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 142.00 - € 412.00
Specifications
| Antigen | Lactoferrin |
|---|---|
| Clone | 103 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019196
![]() |
Invitrogen™
MA552284 |
100 μL |
€ 412.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019310
|
Invitrogen™
MA552285 |
20 μL |
€ 142.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lactoferrin Recombinant Monoclonal antibody specifically detects Lactoferrin in Human samples. It is validated for ELISASpecifications
| Lactoferrin | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; LTF; minisatellite 10r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin | |
| Recombinant Human Lactotransferrin/LTF Protein (Gly 20-Lys710). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LTF |
| 103 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P02788 | |
| 4057 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title