missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoperoxidase Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-61572R
This item is not returnable.
View return policy
Description
Lactoperoxidase Polyclonal specifically detects Lactoperoxidase in Human, Bovine, Primate samples. It is validated for Western Blot.
Specifications
| Lactoperoxidase | |
| Polyclonal | |
| Western Blot | |
| P22079 | |
| LPO | |
| Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-Prot# P22079] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, monkey and bovine. Immunogen has 81% homology to sheep and 73% homology to mouse and rat. | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7 | |
| Rabbit | |
| 100 kDa | |
| 0.1 mL | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 4025 | |
| Human, Bovine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction