missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactose, Monohydrate, Powder, Multi-Compendial, N.F., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
Specifications
Specifications
| CAS | 64044-51-5 |
| Packaging | Poly Bottle |
| Molecular Formula | C12H24O12 |
| Quantity | 2.5 kg |
| Specific Gravity | 1L = 1.53kg |
| Synonym | d-lactose monohydrate, d-+-lactosemonohydrate |
| InChI Key | WSVLPVUVIUVCRA-ZEAJSSQNSA-N |
| SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O |
| IUPAC Name | (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate |
| Molecular Weight (g/mol) | 360.312 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction