missing translation for 'onlineSavingsMsg'
Learn More

LAD1 Mouse anti-Human, Clone: OTI7H8, liquid, TrueMAB™

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA811683

Encompass_Preferred

Product Code. 17545844

  • € 540.87 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Anchoring filament protein which is a component of the basement membrane zone.
Specifications

Specifications

LAD1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00515
LAD1
Human recombit protein fragment corresponding to amino acids 271-517 of human LAD1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI7H8
Unconjugated
LAD1
LadA
Mouse
Affinity Chromatography
RUO
3898
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.