missing translation for 'onlineSavingsMsg'
Learn More

Lafutidine Sulfone, TRC

Product Code. 30464608
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464608 100 mg 100mg
30475445 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464608 Supplier Toronto Research Chemicals Supplier No. TRCL168520100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174583-84-7
Molecular Formula C22H29N3O5S
Molecular Weight (g/mol) 447.55
Synonym 2-[(2-Furanylmethyl)sulfonyl]-N-[(2Z)-4-[[4-(1-piperidinylmethyl)-2-pyridinyl]oxy]-2-buten-1-yl]acetamide, Lafutidine Impurity B
IUPAC Name 2-(furan-2-ylmethylsulfonyl)-N-[(Z)-4-[4-(piperidin-1-ylmethyl)pyridin-2-yl]oxybut-2-enyl]acetamide
SMILES O=C(CS(=O)(=O)Cc1occc1)NC\C=C/COc2cc(CN3CCCCC3)ccn2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H29N3O5S/c26-21(18-31(27,28)17-20-7-6-14-29-20)23-9-2-5-13-30-22-15-19(8-10-24-22)16-25-11-3-1-4-12-25/h2,5-8,10,14-15H,1,3-4,9,11-13,16-18H2,(H,23,26)/b5-2-
Formula Weight 447.1828
Chemical Name or Material Lafutidine Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.