missing translation for 'onlineSavingsMsg'
Learn More

LAG-3 Antibody (11E3), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97662G-0.1ML

Product Code. 18083319

  • € 893.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LAG-3 Monoclonal specifically detects LAG-3 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAG-3
Monoclonal
DyLight 488
50 mM Sodium Borate
LAG3
Recombinant human LAG-3 (lymphocyte activation gene-3).
Adaptive Immunity, Immunology, Innate Immunity
Recognizes the first N-terminal D1 domain of human and monkey LAG-3.
Store at 4°C in the dark.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
11000
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
CD223, CD223 antigen, FDC, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC
Mouse
0.1 mL
3902
Human, Monkey
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.