Learn More
LAG-3 Antibody (631501), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB33281H
Product Code. 18627653
  • € 663.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LAG-3
Monoclonal
HRP
PBS
Rat
0.1 mL
Adaptive Immunity, Cancer, Immunology, Innate Immunity
3902
Store at 4C in the dark.
Flow Cytometry, CyTOF
631501
Flow Cytometry, CyTOF-ready
CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
NS0 mouse myeloma cell line transfected with mouse LAG-3, Ser23-Leu442, Accession # Q61790
RUO
Primary
Mouse
IgG2b