missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 Antibody (874501), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB23193J
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-reported
Specifications
| LAG-3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 874501 | |
| Western Blot, Flow Cytometry, CyTOF-reported | |
| CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
| Mouse myeloma cell line NS0-derived recombinant human LAG-3, Leu23-Leu450, Accession # P18627 | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
| 3902 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction