missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 Antibody (874525), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB23191C
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| LAG-3 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LAG3 | |
| Mouse myeloma cell line NS0-derived partial recombinant human LAG-3 protein (amino acids 23-450) | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Innate Immunity | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry | |
| 874525 | |
| Western Blot, Flow Cytometry | |
| CD223, CD223 antigen, FDC, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction