missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAG-3 Antibody (C9B7W), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-63601AF750
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Mouse samples. It is validated for Flow Cytometry
Specifications
| LAG-3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
| This LAG-3 Antibody (C9B7W) was prepared from murine CD223 Ig fusion protein | |
| 0.125 mL | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
| 3902 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| C9B7W | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction