missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LAG-3 Antibody (CL14773), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26839-25ul
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Immunohistochemistry
Specifications
| LAG-3 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:500-1:1000 | |
| CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
| This antibody was developed against a Synthetic Peptide of P18627. Actual immunogen sequence is proprietary. | |
| 25 μL | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
| 3902 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Immunohistochemistry | |
| CL14773 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction