missing translation for 'onlineSavingsMsg'
Learn More

LAIR-1 (CD305) Mouse anti-Human, Brilliant Violet 605, Clone: DX26, BD Optibuild™

Product Code. 15829139
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15829139 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15829139 Supplier BD Biosciences Supplier No. 744559

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The DX26 monoclonal antibody specifically binds to Leukocyte-associated immunoglobulin-like receptor 1 (LAIR-1/ hLAIR1/Leukocyte-associated Ig-like receptor 1) that is encoded by LAIR1, /and also known as, CD305. LAIR-1 is a ñ32 kDa type I transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory (ITIM) motifs. LAIR-1 is expressed on T cells, B cells, NK cells, monocytes, and dendritic cells. LAIR-1 recruits SHP-1 and SHP-2 tyrosine phosphatases upon activation. Crosslinking of the LAIR-1 expressed on T cells or NK cells can result in strong inhibition of cell-mediated cytotoxicity. Although it is structurally related to human killer cell inhibitory receptors, LAIR-1 does not appear to recognize MHC class I molecules and thus represents a novel MHC class I-independent mechanism of NK cell regulation.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAIR-1 (CD305)
Classification Monoclonal
Clone DX26
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD305, LAIR1, hLAIR1, Leukocyte-associated Ig-like receptor 1
Host Species Mouse
Immunogen Human NK Cell Clone
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.