missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAIR1 Antibody (113), Alexa Fluor™ 532, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-43324AF532
This item is not returnable.
View return policy
Description
LAIR1 Monoclonal specifically detects LAIR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| LAIR1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LAIR1 | |
| The immunogen for this antibody was CD305. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry | |
| 113 | |
| Western Blot, Flow Cytometry | |
| CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
| Armenian Hamster | |
| Protein A or G purified | |
| Immunology | |
| 3903.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction