missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAIR1 Antibody (113), DyLight 488, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-43324G
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunoprecipitation
LAIR1 Monoclonal specifically detects LAIR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| LAIR1 | |
| Monoclonal | |
| 1mg/mL | |
| Western Blot, Flow Cytometry | |
| CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
| Armenian Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry | |
| 113 | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LAIR1 | |
| The immunogen for this antibody was CD305. | |
| 0.1 mL | |
| Immunology | |
| 3903 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction