missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAIR2 Antibody (024), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89685F
This item is not returnable.
View return policy
Description
LAIR2 Monoclonal antibody specifically detects LAIR2 in Human samples. It is validated for ELISA
Specifications
| LAIR2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 024 | |
| ELISA | |
| CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human LAIR2 (NP_002279.2; Met 1-Pro 152). | |
| 0.1 mL | |
| Immunology | |
| 3904 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction