missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAIR2 Antibody (319701), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2665L
This item is not returnable.
View return policy
Description
LAIR2 Monoclonal antibody specifically detects LAIR2 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| LAIR2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 319701 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634 | |
| Mouse myeloma cell line NS0-derived recombinant human LAIR2 isoform 1, Gln22-Pro152, Accession # Q6ISS4 | |
| 0.1 mL | |
| Immunology | |
| 3904 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction