missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAIR2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547366
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 25% cross-reactivity with recombinant human LAIR1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the immunoglobulin superfamily. It was identified by its similarity to leukocyte-associated immunoglobulin-like receptor 1, a membrane-bound receptor that modulates innate immune response. The protein encoded by this locus is a soluble receptor that may play roles in both inhibition of collagen-induced platelet aggregation and vessel formation during placental implantation. This gene maps to a region of 19q13.4, termed the leukocyte receptor cluster, which contains 29 genes in the immunoglobulin superfamily. Alternatively spliced transcript variants have been described for this gene.
Specifications
| LAIR2 | |
| Polyclonal | |
| Unconjugated | |
| LAIR2 | |
| CD306; CD306 antigen; LAIR2; LAIR-2; leukocyte associated immunoglobulin like receptor 2; leukocyte-associated Ig-like receptor 2; leukocyte-associated Ig-like receptor-2; leukocyte-associated immunoglobulin-like receptor 2; MGC71634 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 3904 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q6ISS4 | |
| LAIR2 | |
| Mouse myeloma cell line NS0-derived recombinant human LAIR2 Gln22-Pro152. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction