Learn More
LAMA4 Monoclonal Antibody (OTI5G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA805721
Encompass_Preferred
 View more versions of this product
Product Code. 17565713
  • € 540.87 / 100µL
Description
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
Specifications
Specifications
LAMA4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q16363
LAMA4
Human recombit protein fragment corresponding to amino acids 25-270 of human LAMA4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI5G8
Unconjugated
LAMA4
CMD1JJ, LAMA3, LAMA4*-1
Mouse
Affinity Chromatography
RUO
3910
-20° C, Avoid Freeze/Thaw Cycles