missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (1-155-2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-79124DL594
This item is not returnable.
View return policy
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Lambda Light Chain | |
| Monoclonal | |
| DyLight 594 | |
| IGLC, immunoglobulin lambda constant group | |
| Human IgA1 lambda myeloma protein | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, CyTOF | |
| 1-155-2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction