missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (IGL/3886R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 268.00 - € 496.00
Specifications
| Antigen | Lambda Light Chain |
|---|---|
| Clone | IGL/3886R |
| Concentration | 0.2 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623904
|
Novus Biologicals
NBP3-23945-100ug |
100 μg |
€ 496.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631024
|
Novus Biologicals
NBP3-23945-20ug |
20 μg |
€ 268.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for ELISASpecifications
| Lambda Light Chain | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IGLC, immunoglobulin lambda constant group | |
| Recombinant full-length human Lambda Light Chain protein | |
| Primary | |
| Store at 4°C. |
| IGL/3886R | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 3537 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title