missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody [Janelia Fluor™ 669], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-62139JF669
This item is not returnable.
View return policy
Description
Lambda Light Chain Polyclonal antibody specifically detects Lambda Light Chain in Human,Mouse samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Fluorophore-linked immunosorbent assay,Immunocytochemistry/ Immunofluorescence
Specifications
| Lambda Light Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 669 | |
| IGLC, immunoglobulin lambda constant group | |
| Mouse paraproteins with lambda 1 and lambda 2 light chains | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction