missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (LcN-2), Biotin , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11602B
This item is not returnable.
View return policy
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Lambda Light Chain | |
| Monoclonal | |
| Biotin | |
| IGLC, immunoglobulin lambda constant group | |
| Purified human Lambda Light Chain | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| LcN-2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction