Learn More
Lambda Light Chain Antibody (LcN-2), DyLight 594 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11602DL594
Product Code. 18561367
  • € 762.00 / 0.10mL
Description
Lambda Light Chain Monoclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lambda Light Chain
Monoclonal
DyLight 594
IGLC, immunoglobulin lambda constant group
Purified human Lambda Light Chain
0.1 mL
Adaptive Immunity, Immunology
3537
Store at 4°C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
LcN-2
50mM Sodium Borate
Mouse
Protein A or G purified