missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda Light Chain Antibody (SPM559), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34424PCP
This item is not returnable.
View return policy
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.
Specifications
| Lambda Light Chain | |
| Monoclonal | |
| PerCP | |
| IGLC, immunoglobulin lambda constant group | |
| Mouse | |
| 22.5 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3536 | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM559 | |
| Flow Cytometry | |
| IGLC1 | |
| Purified human IgG | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody is specific to lambda light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the lambda light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction