missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lambda5/IGLL1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99452CL1
This item is not returnable.
View return policy
Description
Lambda5/IGLL1 Polyclonal antibody specifically detects Lambda5/IGLL1 in Mouse samples. It is validated for Western Blot
Specifications
| Lambda5/IGLL1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| 14.1, AGM2, CD179 antigen-like family member B, CD179b, CD179b antigen, Ig lambda-5, IGL1, IGL5CD179B, IGLJ14.1, IGLL, IGO, IGVPB, immunoglobulin lambda-like polypeptide 1, Immunoglobulin omega polypeptide, immunoglobulin omega polypeptide chain, immunoglobulin-related 14.1 protein, Immunoglobulin-related protein 14.1, lambda5, Pre-B lymphocyte-specific protein-2, VPREB2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Lambda5/IGLL1 | |
| 0.1 mL | |
| Immunology | |
| 3543 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction