missing translation for 'onlineSavingsMsg'
Learn More

Lambda5/IGLL1 Antibody, Novus Biologicals™

Product Code. 18198531 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18198531 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18198531 Supplier Novus Biologicals Supplier No. NBP216934

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lambda5/IGLL1 Polyclonal specifically detects Lambda5/IGLL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lambda5/IGLL1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. P15814
Gene Alias 14.1, AGM2, CD179 antigen-like family member B, CD179b, CD179b antigen, Ig lambda-5, IGL1, IGL5CD179B, IGLJ14.1, IGLL, IGO, IGVPB, immunoglobulin lambda-like polypeptide 1, Immunoglobulin omega polypeptide, immunoglobulin omega polypeptide chain, immunoglobulin-related 14.1 protein, Immunoglobulin-related protein 14.1, lambda5, Pre-B lymphocyte-specific protein-2, VPREB2
Gene Symbols IGLL1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human IGLL1. The exact sequence is proprietary.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3543
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.