missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin A + C Antibody (WL4G10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.60 - € 480.90
Specifications
| Antigen | Lamin A + C |
|---|---|
| Clone | WL4G10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653668
|
Novus Biologicals
NBP2-59933-0.025mg |
0.025 mg |
€ 214.00 € 201.60 / 0.03mg Save € 12.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618186
|
Novus Biologicals
NBP2-59933 |
0.1 mg |
€ 509.00 € 480.90 / 0.10mg Save € 28.10 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lamin A + C Monoclonal antibody specifically detects Lamin A + C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Lamin A + C | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Protein Phosphatase, Signal Transduction, Stem Cells | |
| PBS | |
| 4000 | |
| IgG1 κ | |
| Protein A purified |
| WL4G10 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 | |
| GST fused to C-terminal fragment of human lamin A (amino acids residues 422-664) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title