missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B Receptor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 280.00 - € 642.00
Specifications
| Antigen | Lamin B Receptor |
|---|---|
| Dilution | Western Blot, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18467781
|
Novus Biologicals
NBP2-14185-25ul |
25ul |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035085
|
Novus Biologicals
NBP2-14185 |
0.1 mL |
€ 642.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lamin B Receptor Polyclonal specifically detects Lamin B Receptor in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Lamin B Receptor | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| DHCR14B, FLJ43126, Integral nuclear envelope inner membrane protein, lamin B receptor, lamin-B receptor, LMN2R, MGC9041, PHA | |
| LBR | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Cellular Signaling | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 3930 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: MPSRKFADGEVVRGRWPGSSLYYEVEILSHDSTSQLYTVKYKDGTELELKENDIKPLTSFRQRKGGSTSSS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title