missing translation for 'onlineSavingsMsg'
Learn More

Lamin B1 Rabbit anti-Human, Mouse, Rat, Clone: 5H10, Proteintech

Rabbit Recombinant Antibody

€ 153.47 - € 189.00

Specifications

Antigen Lamin B1
Clone 5H10
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17649806
View Documents
Proteintech
80906-1-RR-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17639806
View Documents
Proteintech
80906-1-RR-100UL
100 μL
€ 189.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lamin B1 antibody apecifically detetects Lamin B1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ELISA.
Specifications

Specifications

Lamin B1
1000 μg/mL
Unconjugated
Rabbit
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
4001
Lamin B1 fusion protein Ag3631
Primary
Store at -20°C. Stable for one year after shipment.
5H10
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Liquid
Human, Mouse, Rat
BC012295
LMNB1
IgG
Protein A purification
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.