missing translation for 'onlineSavingsMsg'
Learn More

Lamin B2 Antibody, Novus Biologicals™

Product Code. p-200058051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18685655 25 μL 25µL
18672728 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18685655 Supplier Novus Biologicals Supplier No. NBP24888225ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lamin B2 Polyclonal antibody specifically detects Lamin B2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin B2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias lamin B2, lamin-B2, LMN2LAMB2, MGC2721
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: DEVNKSAKKREGELTVAQGRVKDLESLFHRSEVELAAALSDKRGLESDVAELRAQLAKAEDGHAVAKKQLEKETLM
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 84823
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.