missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B2 Antibody (OTI2C6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72303AF647
This item is not returnable.
View return policy
Description
Lamin B2 Monoclonal specifically detects Lamin B2 in Human samples. It is validated for Western Blot.
Specifications
| Lamin B2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LMNB2 | |
| Full length recombinant protein of human Lamin B2 (NP_116126) produced in HEK293T cells. | |
| 0.1 mL | |
| 84823.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2C6 | |
| Western Blot | |
| lamin B2, lamin-B2, LMN2LAMB2, MGC2721 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction