Learn More
Brady™ Laminated Polyester Biohazard Warning Signs
Click to view available options
Description:
Oblong
Triangular, 100W x 87 mmH
Triangular, 200W x 173 mmH
Triangular, 50W x 43 mmH
Unit Size:
Each
Specifications
Specifications
| Finish | Gloss |
| Material | Laminated Polyester |
| Adhesive Type | Permanent Acrylic |
| Description | Triangular, 50W x 43 mmH |
| Height (Metric) | 43 mm |
| Width (Metric) | 50 mm |
| Shape | Triangular |