Learn More
Brady™ Laminated Polyester CE Marking Labels
Click to view available options
Size (Metric):
18W x 37 mmH
25 mm dia.
37W x 74 mmH
Unit Size:
Each
Pack of 5
Pack of 7
Specifications
Specifications
| Alternate Part Number | PIC 467-DIA 025-B7541 |
| Size (Metric) | 25 mm dia. |
| Diameter (Metric) | 25 mm |
| Shape | Round |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur