Learn More
Brady™ Laminated Polyester CE Marking Labels
Product Code. 10715545 Shop All Brady Products
Change view
Click to view available options
Size (Metric):
18W x 37 mmH
25 mm dia.
37W x 74 mmH
Unit Size:
Each
Pack of 5
Pack of 7
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Unit Size
10715545 37W x 74 mmH Pack of 5
10507434 25 mm dia. Pack of 7
10705545 18W x 37 mmH Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10715545 Supplier Brady™ Supplier No. 250570
  • ONLINE EXCLUSIVE PRICE
    € 6.75
    € 6.06 / Pack of 5
    Save € 0.69
    10% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Alternate Part Number PIC 467-074X037-B7541
Size (Metric) 37W x 74 mmH
Shape Rectangular
Dimensions (L x W) 37W x 74 mmH