Learn More
Brady™ Laminated Polyester Escape Route Signs
Product Code. 10514525
Change view
Click to view available options
Dimensions (L x W):
148 mm W x 297 mm H
148 mm W x 297mm H
148mm W x 297 mm H
148mm W x 297mm H
3.9 x 3.9 in.
5.8 x 11.7 in.
7.9 x 7.9 in.
8.3 x 4.1 in.
Displayed Text:
Direction arrow
Emergency (Left and Down)
Emergency (Left and Up)
Emergency (RIght and Up)
Emergency (Right and Down)
Escape (emergency stairs)
Escape route to the left
Escape route to the right
Rescue ladder
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Displayed Text unitSize
10514525 148 mm W x 297mm H Emergency (Left and Down) Each
10385614 7.9 x 7.9 in. Direction arrow Each
10214994 148 mm W x 297mm H Emergency (Left and Up) Each
10080564 3.9 x 3.9 in. Direction arrow Each
10284894 5.8 x 11.7 in. Emergency (Right and Down) Each
10192504 5.8 x 11.7 in. Emergency (RIght and Up) Each
10336204 148 mm W x 297 mm H Escape (emergency stairs) Each
10030854 148mm W x 297 mm H Escape (emergency stairs) Each
10615115 8.3 x 4.1 in. Escape route to the left Each
10020854 148mm W x 297 mm H Escape route to the left Each
10644155 8.3 x 4.1 in. Escape route to the right Each
10527044 148mm W x 297mm H Escape route to the right Each
10481254 148mm W x 297mm H Rescue ladder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10514525 Supplier Brady™ Supplier No. 250430
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 10-08-2026
Specifications
Certifications/Compliance EU Directive 2002/95/EC
Alternate Part Number PIC 361-297X148-B7541 (BRANDED)
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 297 mm
Thickness 0.024 mm
Width (Metric) 148 mm
Temperature Range (Metric) -40°C to 110°C
Shape Oblong
Dimensions (L x W) 148 mm W x 297mm H
Displayed Text Emergency (Left and Down)
Product Type Escape Route Sign
Color White on Green