Learn More
Brady™ Laminated Polyester Extinguisher Signs: No Water Pictogram
€ 3.04 - € 4.10
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10565694
|
Brady™
250008 |
400 mm |
€ 4.10
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10112314
|
Brady™
250007 |
315 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||