Learn More
Brady™ Laminated Polyester Fire Safety Signs: Fire Hose Pictogram
€ 3.18 - € 4.75
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10254794
|
Brady™
250505 |
200 x 200 mm |
€ 4.75
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10613005
|
Brady™
250503 |
100 x 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||